Assay Details
| Gene Symbol: MRPL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014078.5 | NP_054797.2 | 3 - 4 | 553 | 146 | |
| GenBank mRNA | AB049640.1 | - | 3 - 4 | 279 | 146 | |
| AF112214.1 | - | 3 - 4 | 282 | 146 | ||
| AK311924.1 | - | 3 - 4 | 305 | 146 | ||
| BC009190.2 | - | 3 - 4 | 284 | 3956315 | 146 | |
| BC021744.2 | - | 3 - 4 | 532 | 5260243 | 146 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001564.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020595.hg.1 | ENST00000306185.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020595.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020595.hg.1 | TC08001564.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000521648.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000522717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | ENST00000613356.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011633.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000306185.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000518696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000518918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | ENST00000520677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | NM_014078.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179001.hg.1 | TC0800011633.hg.1 | Clariom D TC |

MRPL13
Genomic Map