| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
ENST00000239849.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
ENST00000358545.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
ENST00000398795.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
ENST00000405262.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
ENST00000544862.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001813.hg.1 |
TC13000154.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006994.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000239849.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000358545.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000398795.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000405262.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
ENST00000544862.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
NM_003701.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
NM_033012.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300147508.hg.1 |
TC1300006994.hg.1 |
Clariom D TC |