Assay Details
| Gene Symbol: MAN1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006699.3 | NP_006690.1 | 8 - 9 | 1880 | 114 | |
| XM_006710302.2 | XP_006710365.1 | 8 - 9 | 1888 | 114 | ||
| XM_011540536.2 | XP_011538838.1 | 8 - 9 | 1950 | 114 | ||
| GenBank mRNA | AF027156.1 | - | 8 - 9 | 1679 | 114 | |
| AK001970.1 | - | 4 - 5 | 357 | 114 | ||
| BC063300.1 | - | 8 - 9 | 1878 | 5263292 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001029.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01016331.hg.1 | ENST00000329466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016331.hg.1 | ENST00000356554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016331.hg.1 | ENST00000369450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016331.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016331.hg.1 | TC01001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016332.hg.1 | ENST00000329466.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016332.hg.1 | ENST00000356554.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016332.hg.1 | ENST00000369450.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016332.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01016332.hg.1 | TC01001029.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168905.hg.1 | TC0100009568.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100168904.hg.1 | TC0100009568.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000356554.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000421535.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000422329.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000449370.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | ENST00000482811.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009568.hg.1 | NM_006699.4 | Refseq |

MAN1A2
Genomic Map