Assay Details
| Gene Symbol: ADAMTS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006988.4 | NP_008919.3 | 3 - 4 | 1663 | 81 | |
| GenBank mRNA | AB037767.1 | - | 3 - 4 | 1323 | 81 | |
| AF060152.1 | - | 3 - 4 | 1394 | 81 | ||
| AF170084.1 | - | 3 - 4 | 1450 | 81 | ||
| AF207664.1 | - | 3 - 4 | 1650 | 81 | ||
| AK314475.1 | - | 3 - 4 | 1663 | 81 | ||
| BC036515.2 | - | 3 - 4 | 1663 | 5266513 | 81 | |
| BX647388.1 | - | 3 - 4 | 1662 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000345.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ovulation from ovarian follicle
kidney development
proteolysis
integrin-mediated signaling pathway
negative regulation of cell proliferation
positive regulation of neuron projection development
tooth eruption
heart trabecula formation
cellular response to vitamin D
cellular response to parathyroid hormone stimulus
cellular response to prostaglandin E stimulus
metalloendopeptidase activity
heparin binding
metallopeptidase activity
zinc ion binding
ovulation from ovarian follicle
kidney development
proteolysis
integrin-mediated signaling pathway
negative regulation of cell proliferation
positive regulation of neuron projection development
tooth eruption
heart trabecula formation
cellular response to vitamin D
cellular response to parathyroid hormone stimulus
cellular response to prostaglandin E stimulus
metalloendopeptidase activity
heparin binding
metallopeptidase activity
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003910.hg.1 | ENST00000284984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003910.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003910.hg.1 | TC21000345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003909.hg.1 | ENST00000284984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003909.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003909.hg.1 | TC21000345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003908.hg.1 | ENST00000284984.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003908.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003908.hg.1 | TC21000345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000492656.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517452.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007821.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | TC2100007821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152720.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | TC2100007821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152719.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | NM_006988.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | TC2100007821.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000284984.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000451462.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000464589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | ENST00000517777.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152721.hg.1 | NM_006988.4 | Refseq |

ADAMTS1
Genomic Map