| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000313070.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000334010.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000360097.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000412584.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000420900.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000449022.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
ENST00000545465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
NM_001009571.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
NM_001167940.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
NM_017954.10 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07028083.hg.1 |
TC07001813.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000313070.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000334010.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000397721.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000412584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000449022.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000462699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000615869.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_001009571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_001167940.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_017954.10 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000313070.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000334010.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000397721.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000412584.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000449022.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000462699.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
ENST00000615869.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_001009571.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_001167940.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
NM_017954.10 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700189608.hg.1 |
TC0700012444.hg.1 |
Clariom D TC |