| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009402.hg.1 |
ENST00000266736.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009402.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12009402.hg.1 |
TC12000743.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200162497.hg.1 |
TC1200008526.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000266736.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000548310.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000549171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
ENST00000553023.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008526.hg.1 |
NM_152435.2 |
Refseq |