| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011782.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011782.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011782.hg.1 |
TC20000861.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011783.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011783.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011783.hg.1 |
TC20000861.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011778.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011778.hg.1 |
ENST00000372970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011778.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011778.hg.1 |
TC20000861.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011784.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20011784.hg.1 |
TC20000861.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
TC2000009180.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164178.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
TC2000009180.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164179.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164184.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164184.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164184.hg.1 |
TC2000009180.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164184.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164184.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164185.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164185.hg.1 |
TC2000009180.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164185.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
ENST00000372970.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009180.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164183.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164183.hg.1 |
NM_016470.7 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000164183.hg.1 |
TC2000009180.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000164183.hg.1 |
ENST00000255174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000164183.hg.1 |
NM_016470.7 |
Refseq |