| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008705.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008705.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008705.hg.1 |
TC13000754.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008706.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008706.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13008706.hg.1 |
TC13000754.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300162572.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162572.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162572.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162572.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162572.hg.1 |
TC1300009321.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300162573.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162573.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162573.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162573.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162573.hg.1 |
TC1300009321.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300162571.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162571.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162571.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300162571.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300162571.hg.1 |
TC1300009321.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
ENST00000282003.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009321.hg.1 |
NM_024546.3 |
Refseq |