Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017855.3 | NP_060325.3 | 3 - 4 | 164 | 150 | |
| XM_005265698.3 | XP_005265755.1 | 4 - 5 | 228 | 150 | ||
| XM_006714245.3 | XP_006714308.1 | 4 - 5 | 205 | 150 | ||
| XM_017008331.1 | XP_016863820.1 | 2 - 3 | 120 | 108 | ||
| GenBank mRNA | BG181363.1 | - | 2 - 3 | 122 | 108 | |
| BG194365.1 | - | 3 - 4 | 167 | 150 | ||
| EF113908.1 | - | 3 - 4 | 166 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

ODAM
Genomic Map