Assay Details
| Gene Symbol: CCDC158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832445.1 | - | 7 - 8 | 2694 | 73 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000388914.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000434846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504667.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000504868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | ENST00000509851.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011061.hg.1 | NM_001042784.1 | Refseq |

CCDC158
Genomic Map