Assay Details
| Gene Symbol: ISL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002202.2 | NP_002193.2 | 2 - 3 | 766 | 76 | |
| GenBank mRNA | AK290742.1 | - | 2 - 3 | 484 | 76 | |
| BC031213.1 | - | 2 - 3 | 767 | 5269369 | 76 | |
| S70721.1 | - | 2 - 3 | 223 | 76 | ||
| U07559.1 | - | 2 - 3 | 457 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
secondary heart field specification
outflow tract septum morphogenesis
outflow tract morphogenesis
endocardial cushion morphogenesis
cardiac right ventricle morphogenesis
regulation of secondary heart field cardioblast proliferation
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
positive regulation of vascular endothelial growth factor production
spinal cord motor neuron cell fate specification
spinal cord motor neuron differentiation
visceral motor neuron differentiation
trigeminal nerve development
pituitary gland development
pancreas development
axon regeneration
retinal ganglion cell axon guidance
positive regulation of insulin secretion
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-1 alpha production
positive regulation of interleukin-1 beta production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
negative regulation of intracellular estrogen receptor signaling pathway
positive regulation of histone acetylation
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of DNA binding
negative regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
mesenchymal cell differentiation
sensory system development
peripheral nervous system neuron axonogenesis
negative regulation of inflammatory response
ventricular cardiac muscle tissue morphogenesis
pharyngeal system development
cardiac muscle cell myoblast differentiation
innervation
atrial septum morphogenesis
cardiac cell fate determination
cellular response to glucocorticoid stimulus
positive regulation of granulocyte colony-stimulating factor production
negative regulation of protein homodimerization activity
negative regulation of canonical Wnt signaling pathway
positive regulation of macrophage colony-stimulating factor production
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
enhancer sequence-specific DNA binding
chromatin binding
protein binding
zinc ion binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
bHLH transcription factor binding
negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
secondary heart field specification
outflow tract septum morphogenesis
outflow tract morphogenesis
endocardial cushion morphogenesis
cardiac right ventricle morphogenesis
regulation of secondary heart field cardioblast proliferation
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
positive regulation of vascular endothelial growth factor production
spinal cord motor neuron cell fate specification
spinal cord motor neuron differentiation
visceral motor neuron differentiation
trigeminal nerve development
pituitary gland development
pancreas development
axon regeneration
retinal ganglion cell axon guidance
positive regulation of insulin secretion
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-1 alpha production
positive regulation of interleukin-1 beta production
positive regulation of interleukin-12 production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
negative regulation of intracellular estrogen receptor signaling pathway
positive regulation of histone acetylation
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of DNA binding
negative regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
neuron fate specification
mesenchymal cell differentiation
sensory system development
peripheral nervous system neuron axonogenesis
negative regulation of inflammatory response
ventricular cardiac muscle tissue morphogenesis
pharyngeal system development
cardiac muscle cell myoblast differentiation
innervation
atrial septum morphogenesis
cardiac cell fate determination
cellular response to glucocorticoid stimulus
positive regulation of granulocyte colony-stimulating factor production
negative regulation of protein homodimerization activity
negative regulation of canonical Wnt signaling pathway
positive regulation of macrophage colony-stimulating factor production
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
enhancer sequence-specific DNA binding
chromatin binding
protein binding
zinc ion binding
ligand-dependent nuclear receptor binding
estrogen receptor binding
bHLH transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05002583.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002583.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002583.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002584.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002584.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002584.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002582.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002582.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002582.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002585.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002585.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002585.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002585.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002586.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002586.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002586.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002586.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002589.hg.1 | ENST00000230658.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002589.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002589.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002589.hg.1 | TC05000212.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007349.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149644.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149644.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149644.hg.1 | TC0500007349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000505475.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149649.hg.1 | TC0500007349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149647.hg.1 | TC0500007349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | ENST00000511384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149646.hg.1 | TC0500007349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149645.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149645.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149645.hg.1 | ENST00000230658.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149645.hg.1 | NM_002202.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149645.hg.1 | TC0500007349.hg.1 | Clariom D TC |

ISL1
Genomic Map