| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022830.hg.1 |
ENST00000344774.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022830.hg.1 |
ENST00000388932.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022830.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022830.hg.1 |
TC09001767.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182958.hg.1 |
TC0900012048.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000344774.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000471784.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
ENST00000484720.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012048.hg.1 |
NM_001001710.2 |
Refseq |