Assay Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012405.3 | NP_036537.1 | 1 - 2 | 269 | 67 | |
GenBank mRNA | AF064084.1 | - | 1 - 2 | 239 | 67 | |
AK092890.1 | - | 2 - 3 | 210 | 67 | ||
BC017037.1 | - | 1 - 2 | 257 | 67 | ||
BC028168.1 | - | 1 - 2 | 225 | 5245125 | 67 | |
CN343480.1 | - | 1 - 2 | 292 | 67 | ||
CR541671.1 | - | 1 - 2 | 196 | 67 | ||
CR541711.1 | - | 1 - 2 | 196 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002147.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK092890.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
C-terminal protein methylation
protein targeting to membrane
post-translational protein modification
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
protein C-terminal carboxyl O-methyltransferase activity
protein C-terminal S-isoprenylcysteine carboxyl O-methyltransferase activity
cellular protein modification process
C-terminal protein methylation
protein targeting to membrane
post-translational protein modification
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
protein C-terminal carboxyl O-methyltransferase activity
protein C-terminal S-isoprenylcysteine carboxyl O-methyltransferase activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | TC01002147.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | ENST00000343813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | ENST00000362035.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | ENST00000489498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | ENST00000535068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033095.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | TC01002147.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | ENST00000343813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | ENST00000362035.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | ENST00000489498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | ENST00000535068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033094.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | TC01002147.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | ENST00000343813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | ENST00000362035.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | ENST00000489498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | ENST00000535068.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033097.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000495791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | ENST00000495791.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01033096.hg.1 | TC01002147.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01033096.hg.1 | ENST00000343813.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033096.hg.1 | ENST00000362035.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033096.hg.1 | ENST00000489498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01033096.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193839.hg.1 | TC0100012673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193840.hg.1 | TC0100012673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193841.hg.1 | TC0100012673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100193842.hg.1 | TC0100012673.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000343813.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000474756.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000489498.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000495791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | ENST00000495791.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012673.hg.1 | NM_012405.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 108793 | - | - |
