| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033090.hg.1 |
ENST00000343813.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033090.hg.1 |
ENST00000489498.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033090.hg.1 |
ENST00000535068.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033090.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01033090.hg.1 |
TC01002147.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000495791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
ENST00000495791.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100193836.hg.1 |
TC0100012673.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000343813.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000474756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000489498.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000495791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
ENST00000495791.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012673.hg.1 |
NM_012405.3 |
Refseq |