Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182854.3 | NP_878274.1 | 3 - 4 | 458 | 64 | |
| GenBank mRNA | AY302441.1 | - | 3 - 4 | 360 | 64 | |
| BC119626.1 | - | 3 - 4 | 311 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001108.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16015510.hg.1 | ENST00000330943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015510.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015510.hg.1 | TC16001108.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015513.hg.1 | ENST00000330943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015513.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015513.hg.1 | TC16001108.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015514.hg.1 | ENST00000330943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015514.hg.1 | ENST00000413750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015514.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015514.hg.1 | TC16001108.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015512.hg.1 | ENST00000330943.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015512.hg.1 | ENST00000413750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015512.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015512.hg.1 | TC16001108.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | TC1600010276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179024.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179023.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179023.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179023.hg.1 | TC1600010276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179023.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179023.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | TC1600010276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179020.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | TC1600010276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179022.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000300590.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000423026.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000568993.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | ENST00000610485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_001144972.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_153337.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010276.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179019.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179019.hg.1 | NM_182854.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600179019.hg.1 | TC1600010276.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600179019.hg.1 | ENST00000330943.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600179019.hg.1 | NM_182854.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 148901 | - | - |

SNX20
Genomic Map