Assay Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030933.2 | NP_112195.2 | 4 - 5 | 1144 | 107 | |
| GenBank mRNA | AF288397.1 | - | 5 - 6 | 802 | 107 | |
| AF288398.1 | - | 4 - 5 | 1143 | 107 | ||
| BC050305.1 | - | 4 - 5 | 1096 | 4837018 | 107 | |
| BC132764.1 | - | 4 - 5 | 1120 | 40146786 | 107 | |
| BC144326.1 | - | 4 - 5 | 1116 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003611.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | ENST00000287709.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | ENST00000367547.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | ENST00000423786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | ENST00000488956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055553.hg.1 | TC01003611.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100226246.hg.1 | TC0100016639.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000367547.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000423786.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000467208.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000483655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000488956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | ENST00000497549.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016639.hg.1 | NM_030933.3 | Refseq |

SHCBP1L
Genomic Map