Assay Details
| Gene Symbol: CIDEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001279.3 | NP_001270.1 | 2 - 3 | 249 | 106 | |
| NM_001318383.1 | NP_001305312.1 | 2 - 3 | 742 | 106 | ||
| GenBank mRNA | AF041378.1 | - | 2 - 3 | 184 | 106 | |
| AY364639.1 | - | 2 - 3 | 184 | 106 | ||
| BC031896.1 | - | 2 - 3 | 314 | 4776542 | 106 | |
| BQ718688.1 | - | 2 - 3 | 197 | 106 | ||
| R73455.1 | - | 2 - 3 | 249 | 106 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000063.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
transcription, DNA-templated
regulation of transcription, DNA-templated
lipid metabolic process
apoptotic process
cell death
positive regulation of sequestering of triglyceride
lipid storage
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of tumor necrosis factor production
response to stilbenoid
regulation of apoptotic process
negative regulation of cytokine secretion
negative regulation of lipid catabolic process
negative regulation of execution phase of apoptosis
regulation of apoptotic DNA fragmentation
protein homodimerization activity
temperature homeostasis
transcription, DNA-templated
regulation of transcription, DNA-templated
lipid metabolic process
apoptotic process
cell death
positive regulation of sequestering of triglyceride
lipid storage
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of tumor necrosis factor production
response to stilbenoid
regulation of apoptotic process
negative regulation of cytokine secretion
negative regulation of lipid catabolic process
negative regulation of execution phase of apoptosis
regulation of apoptotic DNA fragmentation
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000658.hg.1 | ENST00000320477.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000658.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000658.hg.1 | TC18000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000659.hg.1 | ENST00000320477.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000659.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000659.hg.1 | TC18000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000666.hg.1 | ENST00000320477.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000666.hg.1 | ENST00000521296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000666.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000666.hg.1 | TC18000063.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000664.hg.1 | ENST00000320477.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000664.hg.1 | ENST00000521296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000664.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000664.hg.1 | TC18000063.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | TC1800006728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146005.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | TC1800006728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146010.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | TC1800006728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146012.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006728.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146003.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146003.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146003.hg.1 | TC1800006728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146003.hg.1 | ENST00000320477.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146003.hg.1 | NM_001279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000522713.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | TC1800006728.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000520620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000521296.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800146009.hg.1 | ENST00000522713.5 | Ensembl |

CIDEA
Genomic Map