Assay Details
| Gene Symbol: SPCS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021928.3 | NP_068747.1 | 1 - 2 | 278 | 93 | |
| GenBank mRNA | AK026302.1 | - | 1 - 2 | 253 | 93 | |
| AL136660.1 | - | 1 - 2 | 248 | 93 | ||
| AY359044.1 | - | 1 - 2 | 236 | 93 | ||
| BC047290.1 | - | 1 - 2 | 221 | 5224927 | 93 | |
| DB285025.1 | - | 1 - 2 | 280 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000870.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04011971.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011971.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011971.hg.1 | TC04000870.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011970.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011970.hg.1 | TC04000870.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011976.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011976.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011976.hg.1 | TC04000870.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011974.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011974.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04011974.hg.1 | TC04000870.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009379.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164372.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164372.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164372.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164372.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164372.hg.1 | TC0400009379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164375.hg.1 | TC0400009379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164378.hg.1 | TC0400009379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164371.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164371.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164371.hg.1 | TC0400009379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000503362.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000507678.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | ENST00000513139.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | NM_021928.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400164376.hg.1 | TC0400009379.hg.1 | Clariom D TC |

SPCS3
Genomic Map