Assay Details
| Gene Symbol: DUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271762.1 | NP_001258691.1 | 1 - 2 | 411 | 93 | |
| GenBank mRNA | AK000406.1 | - | 1 - 2 | 58 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK000406.1, NM_001271762.1 transcripts.
Set Membership: |
|

DUS2
Genomic Map