Assay Details
| Gene Symbol: ABL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005157.5 | NP_005148.2 | 9 - 10 | 1697 | 71 | |
| NM_007313.2 | NP_009297.2 | 9 - 10 | 2001 | 71 | ||
| GenBank mRNA | AB209456.1 | - | 9 - 10 | 1707 | 71 | |
| AB209642.1 | - | 9 - 10 | 1929 | 71 | ||
| BC117451.1 | - | 9 - 10 | 1627 | 40126002 | 71 | |
| M14752.1 | - | 9 - 10 | 1869 | 71 | ||
| X16416.1 | - | 9 - 10 | 1652 | 71 | ||
More Information
Set Membership: |
|

ABL1
Genomic Map