Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172415.1 | NP_001165886.1 | 1 - 2 | 536 | 56 | |
| NM_004323.5 | NP_004314.5 | 1 - 2 | 536 | 56 | ||
| GenBank mRNA | AF022224.1 | - | 1 - 2 | 494 | 56 | |
| AF116273.1 | - | 1 - 2 | 464 | 56 | ||
| AK096038.1 | - | 2 - 3 | 299 | 56 | ||
| AK222749.1 | - | 1 - 2 | 483 | 56 | ||
| AK316577.1 | - | 1 - 2 | 462 | 56 | ||
| BC001936.1 | - | 1 - 2 | 497 | 2823774 | 56 | |
| BC014774.1 | - | 1 - 2 | 346 | 56 | ||
| BG723775.1 | - | 1 - 2 | 543 | 56 | ||
| KU177932.1 | - | 1 - 2 | 236 | 56 | ||
| KU177933.1 | - | 1 - 2 | 236 | 56 | ||
| U46917.1 | - | 1 - 2 | 496 | 56 | ||
| Z35491.1 | - | 1 - 2 | 514 | 56 | ||
More Information
Set Membership: |
|

BAG1
Genomic Map