| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036404.hg.1 |
ENST00000329601.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036404.hg.1 |
ENST00000330966.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036404.hg.1 |
ENST00000374434.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036404.hg.1 |
NM_152372.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036404.hg.1 |
TC01002357.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100198831.hg.1 |
ENST00000374434.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198831.hg.1 |
NM_152372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198831.hg.1 |
ENST00000374434.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100198831.hg.1 |
NM_152372.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100198831.hg.1 |
TC0100013313.hg.1 |
Clariom D TC |