Assay Details
| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016052.3 | NP_057136.2 | 5 | 5815 | 95 | |
| GenBank mRNA | AB007976.1 | - | 1 | 3663 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRP15| Gene Symbol: RRP15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016052.3 | NP_057136.2 | 5 | 5815 | 95 | |
| GenBank mRNA | AB007976.1 | - | 1 | 3663 | 95 | |
Set Membership: |
|