Assay Details
| Gene Symbol: LOC100291666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF308290.1 | - | 1 | 71 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 28 | LRRC28 | -111.5kb | Anti-sense | Chr. 15 - 99251362 - 99390732 on NCBI Build GRCh38 | |
| Downstream | myocyte enhancer factor 2A | MEF2A |
ADCAD1, RSRFC4, RSRFC9, mef2 |
67.2kb | Anti-sense | Chr. 15 - 99565455 - 99716488 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831986 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373942 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869889 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373947 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373913 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373915 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831992 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373943 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869857 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16831991 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869892 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373877 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869882 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30373861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869865 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869894 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09869849 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P204870 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 39316 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P204870 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P204870 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12348947 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010622.hg.1 | ENST00000623238.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010622.hg.1 | ENST00000623238.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 228880 | - | - |

LOC100291666
Genomic Map