Assay Details
| Gene Symbol: HPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308258.1 | NP_001295187.1 | 1 - 2 | 358 | 82 | |
| GenBank mRNA | AK298478.1 | - | 1 - 2 | 290 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

HPS3
Genomic Map