Assay Details
| Gene Symbol: ILDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199799.1 | NP_001186728.1 | 5 - 6 | 846 | 103 | |
| XM_011512738.2 | XP_011511040.1 | 5 - 6 | 826 | 103 | ||
| GenBank mRNA | AY134857.1 | - | 5 - 6 | 764 | 103 | |
| AY672837.1 | - | 5 - 6 | 688 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001700.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | ENST00000344209.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | ENST00000383663.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | ENST00000393631.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03030770.hg.1 | TC03001700.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300190711.hg.1 | TC0300012178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000273691.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000344209.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000393631.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000460554.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | ENST00000462014.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199799.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_001199800.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012178.hg.1 | NM_175924.3 | Refseq |

ILDR1
Genomic Map