Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017016405.1 | XP_016871894.1 | 1 - 2 | 266 | 74 | |
| GenBank mRNA | BC068471.1 | - | 1 - 2 | 241 | 30343301 | 74 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC068471.1,XM_017016405.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183281.hg.1 | TC1000012164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183282.hg.1 | TC1000012164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000183284.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183284.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183284.hg.1 | TC1000012164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000183280.hg.1 | TC1000012164.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000284690.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000368721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | ENST00000415732.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012164.hg.1 | NM_018180.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 130490 | - | - |

DHX32
Genomic Map