| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023099.hg.1 |
ENST00000284688.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023099.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023099.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10023099.hg.1 |
TC10001740.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000183277.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183277.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183277.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183277.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183277.hg.1 |
TC1000012164.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1000183276.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183276.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183276.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000183276.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000183276.hg.1 |
TC1000012164.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000284690.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000368721.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
ENST00000415732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012164.hg.1 |
NM_018180.2 |
Refseq |