| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007502.hg.1 |
ENST00000371481.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007502.hg.1 |
ENST00000371483.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007502.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007502.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007502.hg.1 |
TC10000665.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000185880.hg.1 |
TC1000012475.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371481.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
ENST00000371483.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_001195755.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012475.hg.1 |
NM_181745.3 |
Refseq |