Assay Details
| Gene Symbol: UBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048241.2 | NP_001041706.1 | 1 - 2 | 105 | 58 | |
| GenBank mRNA | AF313915.1 | - | 1 - 2 | 54 | 58 | |
| AK309258.1 | - | 1 - 2 | 63 | 58 | ||
| AK311889.1 | - | 1 - 2 | 93 | 58 | ||
| BP213482.1 | - | 1 - 2 | 93 | 58 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000164.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF313915.1,AK311889.1,NM_001048241.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19002476.hg.1 | ENST00000358666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002476.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002476.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002476.hg.1 | TC19000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010346.hg.1 | ENST00000583788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002475.hg.1 | ENST00000358666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002475.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002475.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002475.hg.1 | TC19000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007027.hg.1 | ENST00000507979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002471.hg.1 | ENST00000358666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002471.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002471.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002471.hg.1 | TC19000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002472.hg.1 | ENST00000358666.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002472.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002472.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19002472.hg.1 | TC19000164.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700010346.hg.1 | ENST00000583788.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | TC1900006955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149293.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007027.hg.1 | ENST00000507979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000589960.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | ENST00000593087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900006955.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | NM_024292.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | TC1900006955.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000358666.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000586895.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000588141.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000588595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000589372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | ENST00000590068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | NM_001048241.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900149296.hg.1 | NM_024292.3 | Refseq |

UBL5
Genomic Map