| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010856.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010856.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09010856.hg.1 |
TC09000828.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000465773.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
ENST00000484376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009247.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
ENST00000224181.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
ENST00000371634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
NM_000606.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900163176.hg.1 |
TC0900009247.hg.1 |
Clariom D TC |