Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253829.1 | NP_001240758.1 | 7 - 8 | 2307 | 67 | |
| NM_001253830.1 | NP_001240759.1 | 8 - 9 | 2353 | 67 | ||
| NM_152422.4 | NP_689635.3 | 7 - 8 | 2301 | 67 | ||
| NM_177995.2 | NP_818931.1 | 8 - 9 | 2383 | 67 | ||
| XM_005251711.4 | XP_005251768.1 | 8 - 9 | 4325 | 67 | ||
| XM_011518240.2 | XP_011516542.1 | 8 - 9 | 2133 | 67 | ||
| XM_011518241.2 | XP_011516543.1 | 8 - 9 | 2087 | 67 | ||
| XM_017014280.1 | XP_016869769.1 | 9 - 10 | 2192 | 67 | ||
| XM_017014281.1 | XP_016869770.1 | 8 - 9 | 2111 | 67 | ||
| XM_017014282.1 | XP_016869771.1 | 5 - 6 | 2214 | 67 | ||
| GenBank mRNA | AK094631.1 | - | 2 - 3 | 1053 | 67 | |
| AK094773.1 | - | 2 - 3 | 356 | 67 | ||
| AK289692.1 | - | 7 - 8 | 2258 | 67 | ||
| AY171233.1 | - | 8 - 9 | 2388 | 67 | ||
| BC067120.1 | - | 8 - 9 | 2368 | 5539182 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000455.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | ENST00000375360.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004906.hg.1 | TC09000455.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153947.hg.1 | TC0900008010.hg.1 | Clariom D TC |

PTPDC1
Genomic Map