Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014747.2 | NP_055562.2 | 2 - 3 | 440 | 77 | |
| GenBank mRNA | BC003103.1 | - | 2 - 3 | 148 | 3503630 | 77 |
| D87074.1 | - | 2 - 3 | 446 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002558.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC003103.1,D87074.1,NM_014747.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039789.hg.1 | ENST00000372684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039789.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039789.hg.1 | TC01002558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039790.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039790.hg.1 | ENST00000372684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039790.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039790.hg.1 | TC01002558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039788.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039788.hg.1 | ENST00000372684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039788.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039788.hg.1 | TC01002558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039792.hg.1 | ENST00000372684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039792.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039792.hg.1 | TC01002558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039787.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039787.hg.1 | ENST00000372684.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039787.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039787.hg.1 | TC01002558.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203498.hg.1 | TC0100013837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100203500.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203500.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203500.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203500.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203500.hg.1 | TC0100013837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203497.hg.1 | TC0100013837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203503.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203503.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203503.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203503.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203503.hg.1 | TC0100013837.hg.1 | Clariom D TC |

RIMS3
Genomic Map