Assay Details
| Gene Symbol: ANO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031418.3 | NP_113606.2 | 1 - 2 | 185 | 95 | |
| GenBank mRNA | AJ300461.1 | - | 1 - 2 | 892 | 95 | |
| AK302659.1 | - | 1 - 2 | 79 | 95 | ||
| DA247850.1 | - | 1 - 2 | 145 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000275.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302659.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
detection of temperature stimulus
ion transmembrane transport
detection of mechanical stimulus
calcium activated phosphatidylcholine scrambling
calcium activated galactosylceramide scrambling
chloride transmembrane transport
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein dimerization activity
lipid transport
detection of temperature stimulus
ion transmembrane transport
detection of mechanical stimulus
calcium activated phosphatidylcholine scrambling
calcium activated galactosylceramide scrambling
chloride transmembrane transport
intracellular calcium activated chloride channel activity
phospholipid scramblase activity
protein dimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000256737.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000525139.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000529242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000529242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000529242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000529242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000529242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | ENST00000531646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007106.hg.1 | NM_031418.3 | Refseq |

ANO3
Genomic Map