Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC003103.1 | - | 1 - 2 | 46 | 3503630 | 65 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC003103.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203501.hg.1 | TC0100013837.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013837.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | ENST00000372683.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | ENST00000372684.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | NM_014747.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100203499.hg.1 | TC0100013837.hg.1 | Clariom D TC |

RIMS3
Genomic Map