Assay Details
| Gene Symbol: MAN2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002372.3 | NP_002363.2 | 13 - 14 | 3156 | 77 | |
| XM_011543395.2 | XP_011541697.1 | 13 - 14 | 2595 | 77 | ||
| XM_017009472.1 | XP_016864961.1 | 13 - 14 | 2077 | 77 | ||
| GenBank mRNA | AK314090.1 | - | 13 - 14 | 2497 | 77 | |
| BC142656.1 | - | 13 - 14 | 2458 | 40148064 | 77 | |
| BC142696.1 | - | 13 - 14 | 2458 | 40148740 | 77 | |
| D63998.1 | - | 13 - 14 | 2614 | 77 | ||
| U31520.1 | - | 13 - 14 | 2107 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
liver development
mannose metabolic process
N-glycan processing
protein deglycosylation
mitochondrion organization
vacuole organization
respiratory gaseous exchange
lung alveolus development
positive regulation of neurogenesis
retina morphogenesis in camera-type eye
mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase activity
zinc ion binding
hydrolase activity, hydrolyzing N-glycosyl compounds
carbohydrate binding
in utero embryonic development
liver development
mannose metabolic process
N-glycan processing
protein deglycosylation
mitochondrion organization
vacuole organization
respiratory gaseous exchange
lung alveolus development
positive regulation of neurogenesis
retina morphogenesis in camera-type eye
mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase activity
zinc ion binding
hydrolase activity, hydrolyzing N-glycosyl compounds
carbohydrate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000503970.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000505313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | ENST00000513921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008296.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000261483.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000502261.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | ENST00000508043.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | NM_002372.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500156732.hg.1 | TC0500008296.hg.1 | Clariom D TC |

MAN2A1
Genomic Map