Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ409094.1 | - | 1 - 2 | 86 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ409094.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 121636 | - | - |

FAM3B
Genomic Map