Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058186.3 | NP_478066.3 | 6 - 7 | 624 | 79 | |
| NM_206964.1 | NP_996847.1 | 5 - 6 | 480 | 79 | ||
| XM_011529648.2 | XP_011527950.2 | 6 - 7 | 967 | 79 | ||
| XM_011529649.2 | XP_011527951.1 | 6 - 7 | 541 | 79 | ||
| GenBank mRNA | AF375989.1 | - | 6 - 7 | 544 | 79 | |
| AF494379.1 | - | 6 - 7 | 478 | 79 | ||
| AJ409094.1 | - | 7 - 8 | 661 | 79 | ||
| AK125528.1 | - | 6 - 7 | 626 | 79 | ||
| AY040086.1 | - | 5 - 6 | 400 | 79 | ||
| AY358459.1 | - | 6 - 7 | 492 | 79 | ||
| BC057829.1 | - | 6 - 7 | 546 | 30338490 | 79 | |
| BI712971.1 | - | 6 - 7 | 586 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000187.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | ENST00000357985.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | ENST00000398647.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | ENST00000398652.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | ENST00000479810.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21001815.hg.1 | TC21000187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | TC2100007204.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100148553.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000357985.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398646.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398647.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000398652.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000479810.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | ENST00000518236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_058186.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007204.hg.1 | NM_206964.1 | Refseq |

FAM3B
Genomic Map