Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012177.3 | NP_036309.1 | 1 - 2 | 209 | 120 | |
| GenBank mRNA | AF129535.1 | - | 1 - 2 | 161 | 120 | |
| AK314456.1 | - | 1 - 2 | 191 | 120 | ||
| AY079515.1 | - | 1 - 2 | 100 | 120 | ||
| BC018905.1 | - | 1 - 2 | 151 | 3960052 | 120 | |
| BG944882.1 | - | 1 - 2 | 181 | 120 | ||
| DB122061.1 | - | 1 - 2 | 209 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06002240.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
oocyte maturation
spindle assembly involved in female meiosis I
mitotic nuclear division
regulation of mitotic nuclear division
regulation of mitotic cell cycle
vesicle organization
negative regulation of meiotic nuclear division
microtubule polymerization
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein binding
protein kinase binding
metal ion binding
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
oocyte maturation
spindle assembly involved in female meiosis I
mitotic nuclear division
regulation of mitotic nuclear division
regulation of mitotic cell cycle
vesicle organization
negative regulation of meiotic nuclear division
microtubule polymerization
cell division
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
protein binding
protein kinase binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06028166.hg.1 | ENST00000229758.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028166.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028166.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028166.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028166.hg.1 | TC06002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028167.hg.1 | ENST00000229758.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028167.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028167.hg.1 | TC06002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028169.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06028169.hg.1 | TC06002240.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194042.hg.1 | TC0600013602.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194046.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194046.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194046.hg.1 | TC0600013602.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194044.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194044.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194044.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194044.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194044.hg.1 | TC0600013602.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600194043.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194043.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194043.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194043.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194043.hg.1 | TC0600013602.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | ENST00000477822.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013602.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | ENST00000229758.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | ENST00000367241.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | NM_001142522.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | NM_012177.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194041.hg.1 | TC0600013602.hg.1 | Clariom D TC |

FBXO5
Genomic Map