Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242617.1 | NP_001229546.1 | 1 - 2 | 194 | 71 | |
| NM_018698.4 | NP_061168.2 | 2 - 3 | 284 | 71 | ||
| GenBank mRNA | AF201942.1 | - | 1 - 2 | 161 | 71 | |
| AK023289.1 | - | 2 - 3 | 284 | 71 | ||
| BC014888.1 | - | 1 - 2 | 148 | 71 | ||
| BC120984.1 | - | 2 - 3 | 273 | 40116082 | 71 | |
| BC120985.1 | - | 2 - 3 | 273 | 40116088 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000536.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF201942.1,NM_001242617.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007209.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007209.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007207.hg.1 | ENST00000218004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007207.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007207.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | ENST00000218004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | ENST00000372106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | ENST00000372107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007211.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007210.hg.1 | ENST00000372106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007210.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007210.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155412.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155412.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155412.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155412.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155412.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155413.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155409.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155409.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155409.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155409.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155409.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155408.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155408.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155408.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155408.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155408.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155411.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155411.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155411.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 213822 | - | - |

NXT2
Genomic Map