| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025708.hg.1 |
ENST00000228841.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025708.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025708.hg.1 |
TC12001969.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025709.hg.1 |
ENST00000228841.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025709.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025709.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025709.hg.1 |
TC12001969.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025711.hg.1 |
ENST00000228841.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025711.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025711.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025711.hg.1 |
TC12001969.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025706.hg.1 |
ENST00000228841.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025706.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025706.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025706.hg.1 |
TC12001969.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200192651.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192651.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192651.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192651.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192651.hg.1 |
TC1200011920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192655.hg.1 |
TC1200011920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192653.hg.1 |
TC1200011920.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000546404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011920.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000228841.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000548438.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
ENST00000549029.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
NM_000432.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192649.hg.1 |
TC1200011920.hg.1 |
Clariom D TC |