Assay Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171993.1 | NP_001165464.1 | 6 - 7 | 716 | 86 | |
| NM_002150.2 | NP_002141.1 | 4 - 5 | 240 | 86 | ||
| GenBank mRNA | AK057510.1 | - | 6 - 7 | 716 | 86 | |
| AK290826.1 | - | 4 - 5 | 236 | 86 | ||
| BC024287.1 | - | 4 - 5 | 222 | 5087393 | 86 | |
| CD675915.1 | - | 4 - 5 | 369 | 86 | ||
| X72389.1 | - | 4 - 5 | 225 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002066.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12027179.hg.1 | ENST00000543163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027179.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027179.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027179.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027180.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027180.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027180.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | ENST00000289004.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | ENST00000543163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027169.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027178.hg.1 | ENST00000543163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027178.hg.1 | ENST00000545969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027178.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027178.hg.1 | TC12002066.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012215.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195490.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195490.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195490.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195490.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195490.hg.1 | TC1200012215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195491.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195491.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195491.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195491.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195491.hg.1 | TC1200012215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000289004.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000535114.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000542159.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | ENST00000543163.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | NM_002150.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195479.hg.1 | TC1200012215.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195492.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195492.hg.1 | NM_001171993.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195492.hg.1 | TC1200012215.hg.1 | Clariom D TC |

HPD
Genomic Map