| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000590.hg.1 |
ENST00000381434.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000590.hg.1 |
ENST00000456383.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000590.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000590.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09000590.hg.1 |
TC09000045.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000463336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000463336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000417746.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000463336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000463336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_001199641.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900006577.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000381434.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000456383.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
ENST00000611532.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
NM_001199640.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
NM_033439.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900145025.hg.1 |
TC0900006577.hg.1 |
Clariom D TC |