Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242618.1 | NP_001229547.1 | 1 - 2 | 149 | 77 | |
| GenBank mRNA | AF212223.1 | - | 2 - 3 | 140 | 77 | |
| AJ278323.1 | - | 1 - 2 | 149 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000536.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF212223.1,NM_001242618.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007212.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007212.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007212.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | ENST00000218004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | ENST00000372106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | ENST00000372107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007213.hg.1 | TC0X000536.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155416.hg.1 | TC0X00008097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000218004.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372106.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | ENST00000372107.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242617.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008097.hg.1 | NM_018698.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155415.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155415.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155415.hg.1 | ENST00000372103.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155415.hg.1 | NM_001242618.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155415.hg.1 | TC0X00008097.hg.1 | Clariom D TC |

NXT2
Genomic Map