Assay Details
| Gene Symbol: ILF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527987.1 | XP_011526289.1 | 3 - 4 | 359 | 84 | |
| XM_017026764.1 | XP_016882253.1 | 3 - 4 | 555 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527987.1, XM_017026764.1 transcripts.
Set Membership: |
|

ILF3
Genomic Map