Assay Details
| Gene Symbol: CLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006831.2 | NP_006822.1 | 2 - 3 | 749 | 73 | |
| GenBank mRNA | AK313007.1 | - | 2 - 3 | 725 | 73 | |
| BC000446.1 | - | 2 - 3 | 737 | 2821321 | 73 | |
| U73524.1 | - | 2 - 3 | 1333 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000470.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA polyadenylation
mRNA cleavage
tRNA splicing, via endonucleolytic cleavage and ligation
phosphorylation
cerebellar cortex development
targeting of mRNA for destruction involved in RNA interference
mRNA 3'-end processing
siRNA loading onto RISC involved in RNA interference
ATP binding
ATP-dependent polydeoxyribonucleotide 5'-hydroxyl-kinase activity
polydeoxyribonucleotide kinase activity
ATP-dependent polyribonucleotide 5'-hydroxyl-kinase activity
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA polyadenylation
mRNA cleavage
tRNA splicing, via endonucleolytic cleavage and ligation
phosphorylation
cerebellar cortex development
targeting of mRNA for destruction involved in RNA interference
mRNA 3'-end processing
siRNA loading onto RISC involved in RNA interference
ATP binding
ATP-dependent polydeoxyribonucleotide 5'-hydroxyl-kinase activity
polydeoxyribonucleotide kinase activity
ATP-dependent polyribonucleotide 5'-hydroxyl-kinase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005674.hg.1 | TC11000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005673.hg.1 | TC11000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005672.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005672.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005672.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005672.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11005672.hg.1 | TC11000470.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200374.hg.1 | TC1100013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200373.hg.1 | TC1100013001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000525602.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | ENST00000533905.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013001.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000302731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000529430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | ENST00000533682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | NM_001142597.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | NM_006831.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200375.hg.1 | TC1100013001.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 114097 | - | - |

CLP1
Genomic Map