| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002212.hg.1 |
TC01000142.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002212.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002212.hg.1 |
ENST00000389575.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01002212.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100147532.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147532.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100147532.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147532.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100147532.hg.1 |
TC0100006857.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000294484.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000304391.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
ENST00000423056.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006857.hg.1 |
NM_020780.1 |
Refseq |