Assay Details
| Gene Symbol: LINC01114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038231.1 | 2 - 3 | 1756 | 75 | ||
| GenBank mRNA | AK090794.1 | - | 1 - 2 | 1432 | 75 | |
| AK096621.1 | - | 2 - 3 | 1756 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002161.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02035183.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035183.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035183.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035183.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035183.hg.1 | TC02002161.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035188.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035188.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035188.hg.1 | TC02002161.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035182.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035182.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035182.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035182.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02035182.hg.1 | TC02002161.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200200645.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200645.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200645.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200645.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200645.hg.1 | TC0200013733.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200200646.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200646.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200646.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200646.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200646.hg.1 | TC0200013733.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013733.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200636.hg.1 | TC0200013733.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000421071.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000424321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | ENST00000425879.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | NR_038231.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200200635.hg.1 | TC0200013733.hg.1 | Clariom D TC |

LINC01114
Genomic Map