Assay Details
| Gene Symbol: LINC01503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120685.1 | 2 - 3 | 193 | 67 | ||
| GenBank mRNA | DA958985.1 | - | 2 - 3 | 193 | 67 | |
| H43169.1 | - | 1 - 2 | 141 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000423122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000427109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008912.hg.1 | NR_120686.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000436710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | ENST00000444125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | NR_120685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900160314.hg.1 | TC0900008912.hg.1 | Clariom D TC |

LINC01503
Genomic Map