| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007655.hg.1 |
ENST00000430648.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007655.hg.1 |
ENST00000566570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007655.hg.1 |
ENST00000566570.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007655.hg.1 |
NR_026929.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007689.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700153147.hg.1 |
TC0700007689.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007284.hg.1 |
ENST00000619819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
ENST00000561575.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
ENST00000563054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
NR_120505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700153146.hg.1 |
TC0700007689.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007655.hg.1 |
ENST00000430648.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007655.hg.1 |
ENST00000566570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007655.hg.1 |
ENST00000566570.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007655.hg.1 |
NR_026929.1 |
Refseq |